ENA432538608 Enamine

1-amino-3-(propan-2-yl)-2,3-dihydro-1H-indene-1-carboxylic acid

  • MDL number MFCD28139505
  • Cas Number : 1803591-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193731
Availability details for
Quantity
  • Availability
  • Add to Cart