ENA432538617 Enamine

ethyl 1,3-thiazolidine-4-carboxylate hydrochloride

  • MDL number MFCD00206007
  • Cas Number : 66223-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-118
Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194029
Availability details for
Quantity
  • Availability
  • Add to Cart