ENA432538621 Enamine

1-(4-cyanophenyl)-3,3-dimethylurea

  • MDL number MFCD06154342
  • Cas Number : 82261-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194063
Availability details for
Quantity
  • Availability
  • Add to Cart