ENA432538629 Enamine

4,4-dimethylpent-1-en-3-ol

  • MDL number MFCD21232174
  • Cas Number : 24580-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-194079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194079
Availability details for
Quantity
  • Availability
  • Add to Cart