ENA432538636 Enamine

2,2,2-trifluoroethyl N-(4,6-dimethylpyridin-2-yl)carbamate

  • MDL number MFCD28139513
  • Cas Number : 1712667-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194127
Availability details for
Quantity
  • Availability
  • Add to Cart