ENA432538639 Enamine

4-(4-methyl-3-nitrophenyl)morpholine

  • MDL number MFCD20483669
  • Cas Number : 245117-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194139
Availability details for
Quantity
  • Availability
  • Add to Cart