ENA432538640 Enamine

3-hydroxy-4-iodobenzamide

  • MDL number MFCD11046224
  • Cas Number : 1011713-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194146
Availability details for
Quantity
  • Availability
  • Add to Cart