ENA432538643 Enamine

(1,3-dioxaindan-5-yl)methanethiol

  • MDL number MFCD00068473
  • Cas Number : 34817-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194163

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194163
Availability details for
Quantity
  • Availability
  • Add to Cart