ENA432538645 Enamine

1-(5-amino-2-methylphenyl)-3,3-dimethylurea

  • MDL number MFCD21809471
  • Cas Number : 1514414-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194167
Availability details for
Quantity
  • Availability
  • Add to Cart