ENA432538648 Enamine

2-(butan-2-yl)pyrimidin-5-amine

  • MDL number MFCD16706819
  • Cas Number : 1247151-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194170
Availability details for
Quantity
  • Availability
  • Add to Cart