ENA432538650 Enamine

2-(azetidin-3-yl)acetamide hydrochloride

  • MDL number MFCD28139517
  • Cas Number : 1795493-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194172
Availability details for
Quantity
  • Availability
  • Add to Cart