ENA432538658 Enamine

2-aminoethyl acetate hydrochloride

  • MDL number MFCD28139520
  • Cas Number : 20739-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-194180
Availability details for
Quantity
  • Availability
  • Add to Cart