ENA432538664 Enamine

(2-chloro-4-iodophenyl)methanamine hydrochloride

  • MDL number MFCD28139523
  • Cas Number : 1803605-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194192
Availability details for
Quantity
  • Availability
  • Add to Cart