ENA432538666 Enamine

6-hydroxy-5-(propan-2-yl)-3,4-dihydropyrimidin-4-one

  • MDL number MFCD18843988
  • Cas Number : 90345-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194196
Availability details for
Quantity
  • Availability
  • Add to Cart