ENA432538668 Enamine

3-methyl-2-phenylbutanal

  • MDL number MFCD11934481
  • Cas Number : 2439-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194203

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194203
Availability details for
Quantity
  • Availability
  • Add to Cart