ENA432538673 Enamine

3-(2,6-dichlorophenyl)-5-propyl-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD19092969
  • Cas Number : 774605-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194245
Availability details for
Quantity
  • Availability
  • Add to Cart