ENA432538674 Enamine

3-(3,4-dichlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD07376113
  • Cas Number : 4402-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194247
Availability details for
Quantity
  • Availability
  • Add to Cart