ENA432538675 Enamine

3-(2,6-dichlorophenyl)-5-(propan-2-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD18072452
  • Cas Number : 774605-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194249
Availability details for
Quantity
  • Availability
  • Add to Cart