ENA432538677 Enamine

3-(3-chlorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD07376075
  • Cas Number : 92545-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194253
Availability details for
Quantity
  • Availability
  • Add to Cart