ENA432538681 Enamine

ethyl 4-amino-5-cyano-2-hydroxybenzoate

  • MDL number MFCD00728545
  • Cas Number : 72817-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194261
Availability details for
Quantity
  • Availability
  • Add to Cart