ENA432538687 Enamine

9-(bromomethyl)-9H-fluorene

  • MDL number MFCD12025178
  • Cas Number : 56954-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194284
Availability details for
Quantity
  • Availability
  • Add to Cart