ENA432538688 Enamine

2-methylmorpholine-4-carbothioamide

  • MDL number MFCD09729467
  • Cas Number : 953734-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194292
Availability details for
Quantity
  • Availability
  • Add to Cart