ENA432538691 Enamine

ethyl 1-[3-(aminomethyl)phenyl]-1H-pyrazole-3-carboxylate hydrochloride

  • MDL number MFCD28139526
  • Cas Number : 1795304-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194295
Availability details for
Quantity
  • Availability
  • Add to Cart