ENA432538694 Enamine

tert-butyl N-methyl-N-(prop-2-yn-1-yl)carbamate

  • MDL number MFCD22548360
  • Cas Number : 124045-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194303
Availability details for
Quantity
  • Availability
  • Add to Cart