ENA432538697 Enamine

7-bromo-4,4-dimethyl-1,2,3,4-tetrahydronaphthalen-1-one

  • MDL number MFCD07357378
  • Cas Number : 166978-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-57
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176532
Availability details for
Quantity
  • Availability
  • Add to Cart