ENA432538698 Enamine

2-bromo-5-methyl-4-nitropyridin-1-ium-1-olate

  • MDL number MFCD00234157
  • Cas Number : 60323-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-154
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194323

Safety Information

signalword Warning
pcodes P201,P202,P261,P264,P270,P271,P280,P281,P301+P312,P302+P352,P304+P340,P305+P351+P338,P308+P313,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
hcodes Carc Cat 3,Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-194323
pictograms GHS07,GHS08
hstatements H302,H312,H315,H319,H332,H335,H351
Availability details for
Quantity
  • Availability
  • Add to Cart