ENA432538703 Enamine

2-amino-2-[4-chloro-2-(trifluoromethyl)phenyl]ethan-1-ol hydrochloride

  • MDL number MFCD24658487
  • Cas Number : 1701984-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194330
Availability details for
Quantity
  • Availability
  • Add to Cart