ENA432538708 Enamine

4-hydroxy-1-methyl-1H-pyrazole-3,5-dicarboxylic acid

  • MDL number MFCD28145200
  • Cas Number : 1803602-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177204
Availability details for
Quantity
  • Availability
  • Add to Cart