ENA432538717 Enamine

2-chloro-5H,6H,7H,8H,9H,10H-cycloocta[b]pyridine-3,4-dicarbonitrile

  • MDL number MFCD12033785
  • Cas Number : 1020244-32-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194371
Availability details for
Quantity
  • Availability
  • Add to Cart