ENA432538721 Enamine

2-cyclopropyl-5-iodo-1,3,4-oxadiazole

  • MDL number MFCD28139543
  • Cas Number : 1592357-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194422
Availability details for
Quantity
  • Availability
  • Add to Cart