ENA432538724 Enamine

3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(pyridin-2-yl)propanoic acid

  • MDL number MFCD06740409
  • Cas Number : 886505-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194425
Availability details for
Quantity
  • Availability
  • Add to Cart