ENA432538732 Enamine

2-bromo-4-chloro-5-fluorophenol

  • MDL number MFCD26895908
  • Cas Number : 157165-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194438
Availability details for
Quantity
  • Availability
  • Add to Cart