ENA432538733 Enamine

1-methyl-2-phenylcyclohexan-1-ol

  • MDL number MFCD12067577
  • Cas Number : 94734-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194439
Availability details for
Quantity
  • Availability
  • Add to Cart