ENA432538734 Enamine

3-methylpyrrolidine-3-carboxamide

  • MDL number MFCD18273762
  • Cas Number : 1341559-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194442
Availability details for
Quantity
  • Availability
  • Add to Cart