ENA432538736 Enamine

4-(2,6-dichlorophenyl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD11217966
  • Cas Number : 1096815-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194455
Availability details for
Quantity
  • Availability
  • Add to Cart