ENA432538738 Enamine

2-(1-phenyl-1H-pyrazol-4-yl)ethan-1-ol

  • MDL number MFCD07440132
  • Cas Number : 1020703-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194463
Availability details for
Quantity
  • Availability
  • Add to Cart