ENA432538744 Enamine

2-cycloheptylethan-1-ol

  • MDL number MFCD09926200
  • Cas Number : 4480-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194492
Availability details for
Quantity
  • Availability
  • Add to Cart