ENA432538745 Enamine

sodium 2,3-dihydro-1,4-benzodioxine-6-sulfinate

  • MDL number MFCD24113278
  • Cas Number : 1514973-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194544
Availability details for
Quantity
  • Availability
  • Add to Cart