ENA432538748 Enamine

5-methanesulfonyl-2,3-dimethylaniline

  • MDL number MFCD24627682
  • Cas Number : 1335140-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194547
Availability details for
Quantity
  • Availability
  • Add to Cart