ENA432538756 Enamine

2-methyltetradecan-1-ol

  • MDL number MFCD21671594
  • Cas Number : 38231-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194557
Availability details for
Quantity
  • Availability
  • Add to Cart