ENA432538772 Enamine

1-azabicyclo[2.2.1]heptan-3-one hydrochloride

  • MDL number MFCD21603631
  • Cas Number : 122737-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194585
Availability details for
Quantity
  • Availability
  • Add to Cart