ENA432538775 Enamine

N1-(1-methylcyclopentyl)benzene-1,2-diamine

  • MDL number MFCD14651459
  • Cas Number : 1250505-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194593
Availability details for
Quantity
  • Availability
  • Add to Cart