ENA432538779 Enamine

1-(2-chloropyridin-4-yl)ethan-1-amine

  • MDL number MFCD13188900
  • Cas Number : 1060811-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194598
Availability details for
Quantity
  • Availability
  • Add to Cart