ENA432538783 Enamine

1-[1-(pyridin-2-yl)azetidin-3-yl]piperazine

  • MDL number MFCD21718067
  • Cas Number : 1480740-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194603
Availability details for
Quantity
  • Availability
  • Add to Cart