ENA432538785 Enamine

4-(4H-1,2,4-triazol-4-yl)pyridine

  • MDL number MFCD06250151
  • Cas Number : 25700-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194607
Availability details for
Quantity
  • Availability
  • Add to Cart