ENA432538787 Enamine

2-(benzyloxy)-3,5-dichlorobenzoic acid

  • MDL number MFCD04542750
  • Cas Number : 518978-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194626

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194626
Availability details for
Quantity
  • Availability
  • Add to Cart