ENA432538788 Enamine

5-amino-1-(pyridin-4-yl)-1H-imidazole-4-carboxamide

  • MDL number MFCD28139362
  • Cas Number : 1803605-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179335
Availability details for
Quantity
  • Availability
  • Add to Cart