ENA432538805 Enamine

2-amino-2-[4-bromo-2-(trifluoromethyl)phenyl]ethan-1-ol

  • MDL number MFCD22528882
  • Cas Number : 1391317-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194743
Availability details for
Quantity
  • Availability
  • Add to Cart