ENA432538806 Enamine

7-ethyl-2,6-dioxa-9-azaspiro[4.5]decane

  • MDL number MFCD21127091
  • Cas Number : 1488303-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194744
Availability details for
Quantity
  • Availability
  • Add to Cart