ENA432538809 Enamine

5-chloro-1,2,3-trifluorobenzene

  • MDL number MFCD13194468
  • Cas Number : 36556-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194748
Availability details for
Quantity
  • Availability
  • Add to Cart